Organic reference standard, Oakmoss Extract solution, analytical standard, Supelco®
Catalog # 75264-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1 mg/mL in ethanol
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:90028-68-5
- Description:Oakmoss extract solution
- M-Clarity quality level:MQ100
- Description:Oakmoss Extract solution
- Flash point:14 °C