Order Entry
Slovakia
ContactUsLinkComponent
 
Phytochemical standard, Nobiletin, analytical standard, Supelco®
  92600-10MG
 :  Merck
 :  
undefined
Phytochemical standard, Nobiletin, analytical standard, Supelco®
  92600-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    478-01-3
  • Description:
    Nobiletin, ≥98,0% (HPLC)
  • Formula:
    C₂₁H₂₂O₈
  • Description:
    Nobiletin
  • Form:
    Neat

 

 

Nobiletin suppresses the expression of matrix metalloproteinases 1, 3 and 9, which participate in the break down of the extracellular matrix during tumor metastasis. Nobiletin is also involved in the increased expression of tissue inhibitor of matrix metalloproteinase 1 further preventing extracellular matrix degradation during tumor invasion. This bioflavonoid is believed to achieve both of these functions by disrupting the signal transduction pathway of Ras-Raf-MEK.