Organic reference standard, Nivalenol solution, analytical standard, Supelco®
Catalog # 34131-2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:23282-20-4
- Description:Nivalenol solution
- M-Clarity quality level:MQ100
- Formula:C₁₅H₂₀O₇
- Storage temperature:–20 °C
- Description:Nivalenol solution
Specifications
About this item
Nivalenol is a potential trichothecene mycotoxin, produced by Fusarium nivale.
Genotoxicity of nivalenol is evaluated by alkaline single-cell gel electrophoresis assay.
Nivalenol is a secondary metabolite produced by Fusarium species, which is a plant pathogenic fungus of wheat and other cereals.
Nivalenol was used as standard to investigate in vitro mycotoxin binding of deoxynivalenol and nivalenol by adsorbent materials (activated carbon).