Supelco® Organic Reference Standard, Neosaxitoxin Solution
Catalog # 41619-0.5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:0,5 mL
- Standards:Certified reference material, TraceCERT®
- CAS No.:64296-20-4
- Description:Neosaxitoxin solution
- Storage conditions:–20 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ300
- Synonym(s):neo-Saxitoxin solution; NeoSTX
- Formula:C₁₀H₁₇N₇O₅
- Molecular weight:315,29
- Storage temperature:–20 °C
- Flash point:Not applicable
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.