Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Myricitrin
  67268-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Myricitrin
  67268-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    17912-87-7
  • Description:
    Myricitrin
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00016930
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside; Myricetin 3-O-α-ʟ-rhamnopyranoside; Myricetin 3-O-rhamnoside; Myricitroside
  • Molecular weight:
    464,38