Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Methyldibromoglutaronitrile
Supelco® Organic Reference Standard, Methyldibromoglutaronitrile
Catalog # 34392-500MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Methyldibromoglutaronitrile
Catalog # 34392-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Standards:
    Analytical standard
  • CAS No.:
    35691-65-7
  • Description:
    Methyldibromoglutaronitrile
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H314
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00072483
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,2-Dibromo-2,4-dicyanobutane; 1-Bromo-1-(bromomethyl)-1,3-propanedicarbonitrile; 2-Bromo-2-(bromomethyl)glutaronitrile
  • Melting point:
    51...55 °C
  • Formula:
    C₆H₆Br₂N₂
  • Molecular weight:
    265,93
  • Storage temperature:
    2…8 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

Specifications

About this item

  • GHS signal: GHS05, GHS07
Caution: P260 - P270 - P280 - P301 + P312 - P303 + P361 + P353 - P305 + P351 + P338