Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Methyl 2-octynoate, analytical standard, Supelco®
Organic reference standard, Methyl 2-octynoate, analytical standard, Supelco®
Catalog # 68982-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Methyl 2-octynoate, analytical standard, Supelco®
Catalog # 68982-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    111-12-6
  • Description:
    Methyl 2-octynoate, ≥97,0% (GC)
  • Formula:
    C₉H₁₄O₂
  • Storage temperature:
  • Description:
    Methyl 2-octynoate
  • Boiling point:
    217…220 °C
  • Form:
    Neat

Specifications

About this item