Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, m-Coumaric acid
Supelco® Organic Reference Standard, m-Coumaric acid
Catalog # 92649-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, m-Coumaric acid
Catalog # 92649-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    588-30-7
  • Description:
    m-Coumaric acid
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004390
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-Hydroxycinnamic acid
  • Molecular weight:
    164,16

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338