Order Entry
Slovakia
ContactUsLinkComponent
Phytochemical standard, Matairesinol, analytical standard, Supelco®
Phytochemical standard, Matairesinol, analytical standard, Supelco®
  04157-5MG
 :  Merck
 :  
undefined
Phytochemical standard, Matairesinol, analytical standard, Supelco®
  04157-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    580-72-3
  • Description:
    Matairesinol, ≥95,0% (HPLC)
  • Formula:
    C₂₀H₂₂O₆
  • Storage temperature:
    2…8 °C
  • Description:
    Matairesinol

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.