Supelco® Organic Reference Standard, Lycopene
Catalog # 75051-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:502-65-8
- Description:Lycopene
- Storage conditions:–65...–96 °C
- Shipping conditions:DICE
- MDL No.:MFCD00017350
- M-Clarity quality level:MQ100
- Synonym(s):E160d; ψ,ψ-Carotene; 2,6,10,14,19,23,27,31-Octamethyl-dotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
- Molecular weight:536,87