Supelco® Organic Reference Standard, Luteolin
72511-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:491-70-3
- Description:Luteolin
- Storage conditions:2…8 °C
- Shipping conditions:WICE
- MDL No.:MFCD00017309
- M-Clarity quality level:MQ100
- Synonym(s):3′,4′,5,7-Tetrahydroxyflavone
- Melting point:~330 °C (lit.)
- Molecular weight:286,24