Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Lithol Rubine BK, analytical standard, Supelco®
Organic reference standard, Lithol Rubine BK, analytical standard, Supelco®
Catalog # 90689-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Lithol Rubine BK, analytical standard, Supelco®
Catalog # 90689-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    5281-04-9
  • Description:
    Lithol Rubine BK, ≥95,0% (HPLC)
  • Formula:
    C₁₈H₁₂CaN₂O₆S
  • Description:
    Lithol Rubine BK
  • Form:
    Neat

Specifications

About this item

Lithol rubine BK is a water soluble, synthetic red coloring dye, which may be used in drugs and cosmetics.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.