Supelco® Organic Reference Standard, Lipid Standard, Mono-, Di and Triglyceride Mix
Catalog # 1787-1AMP
Supplier: Merck
CAS Number:
Specifications
- Pk:1 Ampoul
- Standards:Certified reference material, pkg of 40 mg (Neat)
- Description:Lipid Standard, Mono-, Di-, & Triglyceride Mix
- Storage conditions:–10...–25 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Storage temperature:–20 °C
- Pk Info:pkg of 40 mg (Neat)
Specifications
About this item
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.