Supelco® Organic Reference Standard, ʟ-Histidine Hydrochloride Solution
Catalog # 43011-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:645-35-2
- Description:ʟ-Histidine hydrochloride solution
- Storage conditions:2...8 °C
- Hazard class description:H290
- Shipping conditions:STD
- MDL No.:MFCD00064556
- M-Clarity quality level:MQ100
- Synonym(s):ʟ-Histidine monohydrochloride solution
- Formula:C₆H₉N₃O₂.HCl
- Molecular weight:191,62
- Storage temperature:2…8 °C
- Concentration:100 mm amino acid in 0,1 m HCL
Specifications
About this item
Caution: P234 - P390