Order Entry
Slovakia
ContactUsLinkComponent
 
Organic reference standard, Leucomalachite Green-d6, analytical standard, Supelco®
  33944-10MG
 :  Merck
 :  
undefined
Organic reference standard, Leucomalachite Green-d6, analytical standard, Supelco®
  33944-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • Description:
    Leucomalachite Green-d6
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₃D₆H₂₀N₂
  • Description:
    Leucomalachite Green-d6
  • Form:
    Neat