Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Supelco® Organic Reference Standard, ʟ-Amino acids
Supelco® Organic Reference Standard, ʟ-Amino acids
LAA10-1KT
:
Merck
:
Supelco® Organic Reference Standard, ʟ-Amino acids
LAA10-1KT
:
Merck
:
Pk:
1
Standards:
Analytical standard
Description:
ʟ-Amino acids
Storage conditions:
RT
Shipping conditions:
STD
M-Clarity quality level:
MQ200