Supelco® Organic Reference Standard, Keracyanin Chloride
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:18719-76-1
- Storage conditions:–20 °C
- Shipping conditions:WICE
- MDL No.:MFCD00135404
- M-Clarity quality level:MQ100
- Synonym(s):Cyanidin-3-O-rutinoside chloride
- Formula:C₂₇H₃₁ClO₁₆
- Molecular weight:630,98
- Storage temperature:−20 °C
- Flash point:Not applicable
Specifications
Frequently Bought Together