Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Isopentenyl Pyrophosphate Trilithium Salt
  39784-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Isopentenyl Pyrophosphate Trilithium Salt
  39784-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    18687-43-9
  • Description:
    Isopentenyl pyrophosphate trilithium salt
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD17215950
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-Methyl-3-butenyl pyrophosphate trilithium salt; IPP
  • Formula:
    C₅H₉Li₃O₇P₂
  • Molecular weight:
    263,89
  • Flash point:
    Not applicable
  • Form:
    Neat

 

 

  • GHS signal: GHS07
 : Bottomless glass bottle. Contents are inside inserted fused cone.
 : P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338