Phytochemical standard, Hydroxyvalerenic acid, analytical standard, Supelco®
Catalog # 55507-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:1619-16-5
- Description:Hydroxyvalerenic acid, ≥98,0% (HPLC)
- Formula:C₁₅H₂₂O₃
- Storage temperature:2...8 °C
- Description:Hydroxyvalerenic acid
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.