Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Hydrocinnamic acid, analytical standard, Supelco®
Organic reference standard, Hydrocinnamic acid, analytical standard, Supelco®
  80725-100MG
 :  Merck
 :  
undefined
Organic reference standard, Hydrocinnamic acid, analytical standard, Supelco®
  80725-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    501-52-0
  • Description:
    Hydrocinnamic acid, ≥98,5% (HPLC)
  • Melting point:
    45 - 48 °C
  • Description:
    Hydrocinnamic acid
  • Boiling point:
    280 °C
  • Form:
    Neat