Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, HT-2 Toxin solution, analytical standard, Supelco®
Organic reference standard, HT-2 Toxin solution, analytical standard, Supelco®
Catalog # 34136-2ML
Dodávateľ:  Merck
Číslo CAS:  
undefined
Organic reference standard, HT-2 Toxin solution, analytical standard, Supelco®
Catalog # 34136-2ML
Dodávateľ:  Merck
Číslo CAS:  

Špecifikácie

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • Description:
    HT-2 Toxin solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₂H₃₂O₈
  • Storage temperature:
    −20°C
  • Description:
    HT-2 Toxin solution

Špecifikácie

O tomto produkte