Supelco® Organic Reference Standard, Glyceryl tributyrate
73105-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:60-01-5
- Description:Glyceryl tributyrate
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00009392
- M-Clarity quality level:MQ100
- Synonym(s):1,2,3-Tributyrylglycerol; Glycerol tributyrate; Tributyrin
- Formula:(CH₃CH₂CH₂COOCH₂)₂CHOCOCH₂CH₂CH₃
- Molecular weight:302,36
- Density:1,032 g/ml at 20 °C (lit.)
- Flash point:174 °C - Closed cup
- Boiling point:129 - 131 °C/0,03 mmHg (lit.); 287 - 288 °C (lit.)