Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Ginsenoside Rh2
  73658-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Ginsenoside Rh2
  73658-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    78214-33-2
  • Description:
    Ginsenoside Rh2
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00800712
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β,12β)-12,20-Dihydroxydammar-24-en-3-yl-β-ᴅ-glucopyranoside; 20(S)-Ginsenoside-Rh2
  • Molecular weight:
    622,87

 

 

  • GHS signal: GHS07