Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Ginsenoside Rh1
Supelco® Organic Reference Standard, Ginsenoside Rh1
Catalog # 56805-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ginsenoside Rh1
Catalog # 56805-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    63223-86-9
  • Description:
    Ginsenoside Rh1
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD09951797
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl-β-ᴅ-glucopyranoside; Prosapogenin A2; Sanchinoside B2; Sanchinoside Rh1
  • Molecular weight:
    638,87

Specifications

About this item

  • GHS signal: GHS07