Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Ginsenoside Rg5
Supelco® Organic Reference Standard, Ginsenoside Rg5
Catalog # 43016-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ginsenoside Rg5
Catalog # 43016-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    186763-78-0
  • Description:
    Ginsenoside Rg5
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β, 12β,20E)-12-Hydroxydammara-20(22),24-dien-3-yl 2-O-β-ᴅ-glucopyranosyl-β-ᴅ-glucopyranoside
  • Formula:
    C₄₈H₈₂O₁₈
  • Molecular weight:
    767,00
  • Storage temperature:
    2…8 °C
  • Flash point:
    Not applicable

Specifications

About this item

Caution: P264 - P270 - P301 + P312 - P501