Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Ginsenoside Rg3
Supelco® Organic Reference Standard, Ginsenoside Rg3
Catalog # 64139-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ginsenoside Rg3
Catalog # 64139-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    14197-60-5
  • Description:
    Ginsenoside Rg3
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD06410950
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β,12β)-12,20-Dihydroxydammar-24-en-3-yl 2-O-β-ᴅ-glucopyranosyl-β-ᴅ-glucopyranoside; 20(S)-Ginsenoside-Rg3; Rg3
  • Molecular weight:
    785,01

Specifications

About this item

  • GHS signal: GHS07