Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Ginsenoside Rg2
Supelco® Organic Reference Standard, Ginsenoside Rg2
Catalog # 08171-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ginsenoside Rg2
Catalog # 08171-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    52286-74-5
  • Description:
    Ginsenoside Rg2
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00210511
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-(6-deoxy-α-ʟ-mannopyranosyl)-β-ᴅ-glucopyranoside; Chikusetsusaponin I; Panaxoside Rg2; Prosapogenin C2
  • Formula:
    C₄₂H₇₂O₁₃
  • Molecular weight:
    785,01
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P301 + P312 + P330