Supelco® Organic Reference Standard, Ginsenoside Rd
01518-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:52705-93-8
- Description:Ginsenoside Rd
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(3β,12β)-20-(β-ᴅ-Glucopyranosyloxy)-12-hydroxydammar-24-en-3-yl 2-O-β-ᴅ-glucopyranosyl-β-ᴅ-glucopyranoside; Gypenoside VIII
- Formula:C₄₈H₈₂O₁₈
- Molecular weight:947,15
- Storage temperature:2…8 °C
- Flash point:Not applicable
- GHS signal: GHS07
: P264 - P270 - P301 + P312 - P501