Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Ginsenoside Rc
Supelco® Organic Reference Standard, Ginsenoside Rc
Catalog # 44987-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ginsenoside Rc
Catalog # 44987-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    11021-14-0
  • Description:
    Ginsenoside Rc
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00133368
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3β,12β)-20-[(6-O-α-ʟ-Arabinofuranosyl-β-ᴅ-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-β-ᴅ-glucopyranosyl-β-ᴅ-glucopyranoside; NSC 310104; Panaxoside Rc
  • Formula:
    C₅₃H₉₀O₂₂
  • Molecular weight:
    1079,27
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501