Supelco® Organic Reference Standard, Ginsenoside Rb3
Catalog # 42635-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:68406-26-8
- Description:Ginsenoside Rb3
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD10566396
- M-Clarity quality level:MQ100
- Synonym(s):(3β,12β)-3-[(2-O-β-ᴅ-Glucopyranosyl-β-ᴅ-glucopyranosyl)oxy]-12-hydroxydammar-24-en-20-yl 6-O-β-ᴅ-xylopyranosyl-β-ᴅ-glucopyranoside; Gypenoside IV
- Formula:C₅₃H₉₀O₂₂
- Molecular weight:1079,27
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P264 - P270 - P301 + P312 - P501