Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, γ,γ-Dimethylallyl pyrophosphate ammonium salt
Supelco® Organic Reference Standard, γ,γ-Dimethylallyl pyrophosphate ammonium salt
  38426-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, γ,γ-Dimethylallyl pyrophosphate ammonium salt
  38426-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    69382-61-2
  • Description:
    γ,γ-Dimethylallyl pyrophosphate ammonium salt
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-Methyl-2-butenyl pyrophosphate ammonium salt; DMAPP; Prenyl pyrophosphate ammonium salt
  • Formula:
    C₅H₁₂O₇P₂ ·xNH₃
  • Molecular weight:
    246,09 (free acid basis)
  • Flash point:
    Not applicable
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.