Organic reference standard, Gallocatechin solution, analytical standard, Supelco®
Catalog # 49069-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 µg/ml in methanol
- Pk:1
- Standards:Analytical standard
- Description:Gallocatechin solution
- M-Clarity quality level:MQ100
- Storage temperature:−20 °C
- Description:Gallocatechin solution
- Flash point:9 °C
- Pk Info:Ampoule of 0,5 ml