Order Entry
Slovakia
ContactUsLinkComponent
Phytochemical standard, Friedelin, analytical standard, Supelco®
Phytochemical standard, Friedelin, analytical standard, Supelco®
Catalog # 92187-10MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, Friedelin, analytical standard, Supelco®
Catalog # 92187-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    559-74-0
  • Description:
    Friedelin, ≥95,0% (HPLC)
  • Melting point:
    262...265 °C (lit.)
  • Formula:
    C₃₀H₅₀O
  • Description:
    Friedelin
  • Form:
    Neat

Specifications

About this item