Phytochemical standard, Ethyl trans-4-methoxycinnamate, analytical standard, Supelco®
Catalog # 42479-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:24393-56-4
- Description:Ethyl trans-4-methoxycinnamate, ≥95,0% (GC)
- Formula:C₁₂H₁₄O₃
- Description:Ethyl trans-4-methoxycinnamate
- Form:Neat