Phytochemical standard, Eriodictyol, analytical standard, Supelco®
Catalog # 89061-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:552-58-9
- Description:Eriodictyol, ≥98,0% (HPLC)
- Formula:C₁₅H₁₂O₆
- Description:Eriodictyol
- Form:Neat
Specifications
About this item
Eriodictyol is a naturally occurring flavonoid found in citrus fruits. It is isolated from the Chinese herb, Dracocephalum rupestre. It possess antioxidant activity.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.