Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Epigallocatechin solution
  49037-U
 :  Merck
 :  
Supelco® Organic Reference Standard, Epigallocatechin solution
  49037-U
 :  Merck
 :  

 

  • Concentration:
    2000 μg/ml in methanol, ampule of 0,5 ml
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Epigallocatechin solution
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : Epigallocatechin solution is a Certified Reference Material (CRM) produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
 : P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311