Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Epigallocatechin gallate solution
Supelco® Organic Reference Standard, Epigallocatechin gallate solution
Catalog # 49044-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Epigallocatechin gallate solution
Catalog # 49044-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml in methanol
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Epigallocatechin gallate solution
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311