Supelco® Organic Reference Standard, Epicatechin gallate solution
Catalog # 49060-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 μg/ml in methanol, ampule of 0,5 ml
- Pk:1
- Standards:Certified reference material
- Description:Epicatechin gallate solution
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:9 °C
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Certifications: Epicatechin gallate solution is a Certified Reference Material (CRM) produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311