Supelco® Organic Reference Standard, Emodin
Catalog # 30269-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:518-82-1
- Description:Emodin
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00001207
- M-Clarity quality level:MQ100
- Synonym(s):1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione; 1,3,8-Trihydroxy-6-methylanthraquinone; 6-Methyl-1,3,8-trihydroxyanthraquinone; Archin; Emodol; Frangula-emodin; Frangulic acid; Rheum emodin
- Formula:C₁₅H₁₀O₅
- Molecular weight:270,24
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P264 - P270 - P301 + P312 + P330 - P501