Supelco® Organic Reference Standard, Echinacoside
Catalog # 07538-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:82854-37-3
- Description:Echinacoside
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00075695
- M-Clarity quality level:MQ100
- Formula:C₃₅H₄₆O₂₀
- Molecular weight:786,73
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.