Supelco® Organic Reference Standard, DL-Mandelic Acid
41605-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:90-64-2
- Description:ᴅʟ-Mandelic acid
- Storage conditions:RT
- Hazard class description:H318
- Shipping conditions:STD
- MDL No.:MFCD00064250
- M-Clarity quality level:MQ100
- Synonym(s):(±)-α-Hydroxyphenylacetic acid
- Formula:C₆H₅CH(OH)COOH
- Molecular weight:152,15
- Flash point:Not applicable
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.
: P280 - P305 + P351 + P338