Organic reference standard, disperse yellow 9, analytical standard, Supelco®
38464-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:6373-73-5
- Description:Disperse Yellow 9, ≥96,0% (HPLC)
- M-Clarity quality level:MQ100
- Melting point:187...190 °C (lit.)
- Formula:C₁₂H₁₀N₄O₄
- Description:Disperse Yellow 9
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.