Phytochemical standard, Diosmetin, analytical standard, Supelco®
90985-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:520-34-3
- Description:Diosmetin, ≥95,0% (HPLC)
- Formula:C₁₆H₁₂O₆
- Storage temperature:2...8 °C
- Description:Diosmetin
- Form:Neat
It is a flavonoid obtained from Citrus limon. Act as an antioxidant. Inhibits carcinogen activation by inhibiting the CYP1A1 enzyme.
: Bottomless glass bottle. Contents are inside inserted fused cone.