Organic reference standard, Diisobutyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Catalog # 34204-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:358730-88-8
- Description:Diisobutyl phthalate-3,4,5,6-d4
- Formula:C₁₆D₄H₁₈O₄
- Description:Diisobutyl phthalate-3,4,5,6-d4
- Form:Neat