Phytochemical standard, Dihydrokaempferol, analytical standard, Supelco®
Catalog # 92204-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:480-20-6
- Description:Dihydrokaempferol, ≥95,0% (HPLC)
- Formula:C₁₅H₁₂O₆
- Description:Dihydrokaempferol
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.