Organic reference standard, Dihydrocapsaicin, analytical standard, Supelco®
03813-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:19408-84-5
- Description:Dihydrocapsaicin, ≥97,0% (HPLC)
- Formula:C₁₈H₂₉NO₃
- Storage temperature:2...8 °C
- Description:Dihydrocapsaicin