Order Entry
Slovakia
ContactUsLinkComponent
 
Phytochemical standard, Delphinidin chloride, analytical standard, Supelco®
  14389-10MG
 :  Merck
 :  
undefined
Phytochemical standard, Delphinidin chloride, analytical standard, Supelco®
  14389-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    528-53-0
  • Description:
    Delphinidin chloride, ≥90,0% (HPLC)
  • Formula:
    C₁₅H₁₁ClO₇
  • Storage temperature:
    –20 °C
  • Description:
    Delphinidin chloride
  • Form:
    Neat

 

 

It can be seen in anthocyanidin. Antioxidant. Pigment in grapes, cranberries, and pomegranates.