Phytochemical standard, Delphinidin 3-sambubioside chloride, analytical standard, Supelco®
Catalog # 42883-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:53158-73-9
- Description:Delphinidin 3-sambubioside chloride, ≥90,0% (HPLC)
- Formula:C₂₆H₂₉O₁₆Cl
- Storage temperature:–20 °C
- Description:Delphinidin 3-sambubioside chloride