Phytochemical standard, Delphinidin 3-O-beta-D-glucoside chloride, analytical standard, Supelco®
73705-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:6906-38-3
- Description:Delphinidin 3-O-beta-D-glucoside chloride, ≥95,0% (HPLC)
- Formula:C₂₁H₂₁ClO₁₂
- Storage temperature:–20 °C
- Description:Delphinidin 3-O-beta-D-glucoside chloride
- Form:Neat