Phytochemical standard, Delphinidin 3-O-beta-D-galactoside chloride, analytical standard, Supelco®
Catalog # 04301-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:28500-00-7
- Description:Delphinidin 3-O-beta-D-galactoside chloride, ≥90,0% (HPLC)
- Formula:C₂₁H₂₁ClO₁₂
- Storage temperature:–20 °C
- Description:Delphinidin 3-O-beta-D-galactoside chloride